Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 4.96 mm Dia
Catalog No. 5000516477
Encompass
This plug and pin gage is designed for precision measurement and inspection. It features a 4.96 mm diameter and a 2 inch overall length. Made from steel, it is suitable for setting micrometers, verniers, and for no-go inspection.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Ideal for no-go inspection.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-16477 Supplier MSC Supplier No. 40432411
This plug and pin gage is designed for precision measurement and inspection. It features a 4.96 mm diameter and a 2 inch overall length. Made from steel, it is suitable for setting micrometers, verniers, and for no-go inspection.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Ideal for no-go inspection.
  • Includes a certificate of accuracy with NIST traceability.