Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 5.04 mm, 2 in, Steel
Catalog No. 5000532771
Encompass
This Class ZZ Minus Plug & Pin Gage, crafted from steel, measures 5.04 mm in diameter and 2 in overall length. Its black oxide treatment prevents rust, corrosion, and offers visual wear indication, extending its useful life. It's ideal for setting micrometers, NoGo inspections, and calibrating various measuring devices.
  • Manufactured from tool steel with a hardness of 60/65 Rc.
  • Features a black oxide treatment that prevents rust and corrosion.
  • Provides visible wear indication for easier calibration or replacement.
  • Maintains a Class ZZ tolerance of 0.005 mm.
  • Finished to 10 microinch or better.
  • Round within 0.0025 mm.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers, NoGo inspection, and other measuring devices.
Catalog No. 50-005-32771 Supplier MSC Supplier No. 40432494
This Class ZZ Minus Plug & Pin Gage, crafted from steel, measures 5.04 mm in diameter and 2 in overall length. Its black oxide treatment prevents rust, corrosion, and offers visual wear indication, extending its useful life. It's ideal for setting micrometers, NoGo inspections, and calibrating various measuring devices.
  • Manufactured from tool steel with a hardness of 60/65 Rc.
  • Features a black oxide treatment that prevents rust and corrosion.
  • Provides visible wear indication for easier calibration or replacement.
  • Maintains a Class ZZ tolerance of 0.005 mm.
  • Finished to 10 microinch or better.
  • Round within 0.0025 mm.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers, NoGo inspection, and other measuring devices.