Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 5.32 mm, 2 in OAL, Steel
Catalog No. 5000512533
Encompass
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection. This steel gage features a 5.32 mm diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspections, verniers, and other measuring devices.
  • Designed for setting micrometers, NoGo inspection, and other measuring devices.
  • Constructed from durable steel.
  • Features a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-12533 Supplier MSC Supplier No. 40432775
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection. This steel gage features a 5.32 mm diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspections, verniers, and other measuring devices.
  • Designed for setting micrometers, NoGo inspection, and other measuring devices.
  • Constructed from durable steel.
  • Features a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.