Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 5.51 mm Dia, 2 inch OAL, Steel
Catalog No. 5000533827
Encompass
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a general-purpose tool designed for quick and accurate measurements. It is suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. This gage can be used for Go/NoGo inspection and for sizing with micrometers and other measuring devices, offering extended wear life.
  • Designed for quick and accurate measurements.
  • Ideal for measuring hole sizes and calculating distances between holes.
  • Suitable for Go/NoGo inspection and sizing with micrometers.
  • Constructed from durable steel for extended wear life.
  • Features a 5.51 mm diameter.
  • Has an overall length of 2 inches.
  • Offers a Class ZZ tolerance of 0.00508 mm.
  • Includes a certificate of accuracy with NIST traceability.
  • Finished to 10 microinches or better.
Catalog No. 50-005-33827 Supplier MSC Supplier No. 40432965
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a general-purpose tool designed for quick and accurate measurements. It is suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. This gage can be used for Go/NoGo inspection and for sizing with micrometers and other measuring devices, offering extended wear life.
  • Designed for quick and accurate measurements.
  • Ideal for measuring hole sizes and calculating distances between holes.
  • Suitable for Go/NoGo inspection and sizing with micrometers.
  • Constructed from durable steel for extended wear life.
  • Features a 5.51 mm diameter.
  • Has an overall length of 2 inches.
  • Offers a Class ZZ tolerance of 0.00508 mm.
  • Includes a certificate of accuracy with NIST traceability.
  • Finished to 10 microinches or better.