Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 5.98 mm, 2 in
Catalog No. 5000532262
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring instrument designed for various applications including setting micrometers, NoGo inspection, and other measuring devices. It features a 5.98 mm diameter and a 2-inch overall length. Constructed from steel with a 10 Micro finish, it ensures durability and accuracy with a tolerance of 0.00508 mm. Each gage includes a Certificate of Accuracy with NIST Traceability. Made in the United States.
  • Precision measurement for setting micrometers, NoGo inspection, and other measuring devices.
  • High accuracy with a tolerance of 0.00508 mm and 10 Micro finish.
  • Quality assurance includes a Certificate of Accuracy with NIST Traceability.
  • Durable steel construction for long-lasting performance.
  • Manufactured in the United States.
Catalog No. 50-005-32262 Supplier MSC Supplier No. 40433435
This Class ZZ Minus Plug & Pin Gage is a precision measuring instrument designed for various applications including setting micrometers, NoGo inspection, and other measuring devices. It features a 5.98 mm diameter and a 2-inch overall length. Constructed from steel with a 10 Micro finish, it ensures durability and accuracy with a tolerance of 0.00508 mm. Each gage includes a Certificate of Accuracy with NIST Traceability. Made in the United States.
  • Precision measurement for setting micrometers, NoGo inspection, and other measuring devices.
  • High accuracy with a tolerance of 0.00508 mm and 10 Micro finish.
  • Quality assurance includes a Certificate of Accuracy with NIST Traceability.
  • Durable steel construction for long-lasting performance.
  • Manufactured in the United States.