Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 6.99 mm, 2 in, Steel
Catalog No. 5000527030
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel, it is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Applicable for setting micrometers.
  • Suitable for NoGo inspection.
  • Usable with verniers and other measuring devices.
  • Made from steel for durability.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-27030 Supplier MSC Supplier No. 40434441
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel, it is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Applicable for setting micrometers.
  • Suitable for NoGo inspection.
  • Usable with verniers and other measuring devices.
  • Made from steel for durability.
  • Features a 10 micro coating/finish.