Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 8.03 mm Dia, 2 in OAL
Catalog No. 5000532290
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. Constructed from durable steel, it features an 8.03 mm diameter and a 2 inch overall length, making it ideal for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. Each gage includes a certificate of accuracy with NIST traceability.
  • Constructed from steel
  • Suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish
Catalog No. 50-005-32290 Supplier MSC Supplier No. 40435489
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. Constructed from durable steel, it features an 8.03 mm diameter and a 2 inch overall length, making it ideal for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices. Each gage includes a certificate of accuracy with NIST traceability.
  • Constructed from steel
  • Suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish