Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 8.12 mm, 2 in, Steel
Catalog No. 5000536169
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate calibration and inspection. Made from durable steel, it features an 8.12 mm diameter and a 2 in overall length, making it ideal for setting micrometers and conducting no-go inspections.
  • Diameter of 8.12 mm for precise measurements.
  • Overall length of 2 in for versatility.
  • Tolerance of 0.00508 mm ensures accuracy.
  • Constructed from steel for durability.
  • 10 micro finish for a smooth surface.
  • Suitable for setting micrometers and other measuring devices.
  • Ideal for no-go inspection.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-36169 Supplier MSC Supplier No. 40435570
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate calibration and inspection. Made from durable steel, it features an 8.12 mm diameter and a 2 in overall length, making it ideal for setting micrometers and conducting no-go inspections.
  • Diameter of 8.12 mm for precise measurements.
  • Overall length of 2 in for versatility.
  • Tolerance of 0.00508 mm ensures accuracy.
  • Constructed from steel for durability.
  • 10 micro finish for a smooth surface.
  • Suitable for setting micrometers and other measuring devices.
  • Ideal for no-go inspection.
  • Includes a certificate of accuracy with NIST traceability.