Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 8.20 mm Diameter, 2 in Length
Catalog No. 5000527028
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications. Made from steel with a 10 micro finish, it ensures accurate readings for setting micrometers, verniers, and for no-go inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Features an 8.20 mm diameter of measurement.
  • Has an overall length of 2 in.
  • Provides a tolerance of 0.00508 mm.
  • Constructed from durable steel.
  • Finished with a 10 micro coating.
  • Suitable for setting micrometers and verniers, and for no-go inspection.
Catalog No. 50-005-27028 Supplier MSC Supplier No. 40435653
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications. Made from steel with a 10 micro finish, it ensures accurate readings for setting micrometers, verniers, and for no-go inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Features an 8.20 mm diameter of measurement.
  • Has an overall length of 2 in.
  • Provides a tolerance of 0.00508 mm.
  • Constructed from durable steel.
  • Finished with a 10 micro coating.
  • Suitable for setting micrometers and verniers, and for no-go inspection.