Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 8.46 mm, 10 Microfinish
Catalog No. 5000529183
Encompass
This Class ZZ Minus plug and pin gage offers precise dimensional verification with an 8.46 mm diameter and a tolerance of 0.00508 mm. Constructed from steel and featuring a 10 Microfinish, it measures 2 inches in overall length. It is specifically designed for NoGo inspection and comes with a Certificate of Accuracy, including NIST Traceability.
  • 8.46 mm diameter
  • Class ZZ Minus for precise fit
  • Tolerance of 0.00508 mm
  • Made from durable steel
  • Features a 10 Microfinish
  • 2 inch overall length
  • Ideal for NoGo inspection
  • Includes Certificate of Accuracy with NIST Traceability
Catalog No. 50-005-29183 Supplier MSC Supplier No. 40435919
This Class ZZ Minus plug and pin gage offers precise dimensional verification with an 8.46 mm diameter and a tolerance of 0.00508 mm. Constructed from steel and featuring a 10 Microfinish, it measures 2 inches in overall length. It is specifically designed for NoGo inspection and comes with a Certificate of Accuracy, including NIST Traceability.
  • 8.46 mm diameter
  • Class ZZ Minus for precise fit
  • Tolerance of 0.00508 mm
  • Made from durable steel
  • Features a 10 Microfinish
  • 2 inch overall length
  • Ideal for NoGo inspection
  • Includes Certificate of Accuracy with NIST Traceability