Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 8.80 mm, 2 in
Catalog No. 5000527860
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. Constructed from durable steel with a 10 Micro finish, it provides reliable performance and longevity. It is ideal for tasks such as setting micrometers, conducting NoGo inspections, and calibrating verniers and other measuring devices.
  • Features an 8.80 mm diameter
  • Has a 2 inch overall length
  • Constructed from durable steel
  • Finished with a 10 Micro coating
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
Catalog No. 50-005-27860 Supplier MSC Supplier No. 40436255
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. Constructed from durable steel with a 10 Micro finish, it provides reliable performance and longevity. It is ideal for tasks such as setting micrometers, conducting NoGo inspections, and calibrating verniers and other measuring devices.
  • Features an 8.80 mm diameter
  • Has a 2 inch overall length
  • Constructed from durable steel
  • Finished with a 10 Micro coating
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices