Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 8.98 mm, 2 in OAL, Steel
Catalog No. 5000528910
Encompass
This Class ZZ Minus Plug & Pin Gage has a diameter of 8.98 mm and an overall length of 2 inches. It features a 10 Micro coating/finish and is made of steel. This gage is suitable for applications such as setting micrometers, NoGo inspection, verniers, and other measuring devices. It also includes a Certificate of Accuracy with NIST Traceability.
  • Diameter of 8.98 mm
  • Overall length of 2 inches
  • Tolerance of 0.00508 mm
  • Made from steel
  • Features a 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-28910 Supplier MSC Supplier No. 40436438
This Class ZZ Minus Plug & Pin Gage has a diameter of 8.98 mm and an overall length of 2 inches. It features a 10 Micro coating/finish and is made of steel. This gage is suitable for applications such as setting micrometers, NoGo inspection, verniers, and other measuring devices. It also includes a Certificate of Accuracy with NIST Traceability.
  • Diameter of 8.98 mm
  • Overall length of 2 inches
  • Tolerance of 0.00508 mm
  • Made from steel
  • Features a 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability