Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 9.20 mm, 2 in
Catalog No. 5000532812
Encompass
This Class ZZ Minus Plug & Pin Gage features a 9.20 mm diameter and 2 inch overall length. Constructed from steel with a 10 Micro finish, it is designed for precise measurement applications. Ideal for setting micrometers, verniers, and other measuring devices, it is particularly suited for NoGo inspection.
  • Class ZZ Minus Plug & Pin Gage
  • 9.20 mm diameter
  • 2 inch overall length
  • Steel material
  • 10 Micro finish
  • Certificate of accuracy with NIST traceability included
  • Designed for setting micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
Catalog No. 50-005-32812 Supplier MSC Supplier No. 40436651
This Class ZZ Minus Plug & Pin Gage features a 9.20 mm diameter and 2 inch overall length. Constructed from steel with a 10 Micro finish, it is designed for precise measurement applications. Ideal for setting micrometers, verniers, and other measuring devices, it is particularly suited for NoGo inspection.
  • Class ZZ Minus Plug & Pin Gage
  • 9.20 mm diameter
  • 2 inch overall length
  • Steel material
  • 10 Micro finish
  • Certificate of accuracy with NIST traceability included
  • Designed for setting micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection