Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 9.44 mm Diameter, 2 in Overall Length
Catalog No. 5000528903
Encompass
This Class ZZ Minus Plug & Pin Gage features a 9.44 mm diameter and a 2 in overall length. It is constructed from steel with a 10 Micro coating/finish and offers a tolerance of 0.00508 mm. This gage is supplied with a Certificate of Accuracy with NIST Traceability and is designed for applications such as setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Diameter of measurement: 9.44 mm
  • Overall length: 2 in
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • 10 Micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-28903 Supplier MSC Supplier No. 40436891
This Class ZZ Minus Plug & Pin Gage features a 9.44 mm diameter and a 2 in overall length. It is constructed from steel with a 10 Micro coating/finish and offers a tolerance of 0.00508 mm. This gage is supplied with a Certificate of Accuracy with NIST Traceability and is designed for applications such as setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Diameter of measurement: 9.44 mm
  • Overall length: 2 in
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • 10 Micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability