Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 9.50 mm, 2 in OAL, Steel
Catalog No. 5000529489
Encompass
This Class ZZ Minus Plug & Pin Gage features a 9.50 mm diameter and a 2 inch overall length. Constructed from steel, it has a 10 micro coating/finish. It is designed for precise measurement applications such as setting micrometers, performing NoGo inspections, and use with verniers and other measuring devices.
  • 9.50 mm diameter
  • 2 inch overall length
  • 10 micro coating/finish
  • Steel construction
  • Suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-29489 Supplier MSC Supplier No. 40436958
This Class ZZ Minus Plug & Pin Gage features a 9.50 mm diameter and a 2 inch overall length. Constructed from steel, it has a 10 micro coating/finish. It is designed for precise measurement applications such as setting micrometers, performing NoGo inspections, and use with verniers and other measuring devices.
  • 9.50 mm diameter
  • 2 inch overall length
  • 10 micro coating/finish
  • Steel construction
  • Suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability