Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 9.59 mm, 2 in OAL
Catalog No. 5000516511
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks. Made from steel, it features a 9.59 mm diameter and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspections, and other measuring devices.
  • Includes Certificate of Accuracy with NIST traceability
  • Designed for setting micrometers
  • Suitable for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Constructed from steel
  • Features a 10 Micro coating/finish
Catalog No. 50-005-16511 Supplier MSC Supplier No. 40437048
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks. Made from steel, it features a 9.59 mm diameter and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspections, and other measuring devices.
  • Includes Certificate of Accuracy with NIST traceability
  • Designed for setting micrometers
  • Suitable for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Constructed from steel
  • Features a 10 Micro coating/finish