Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 9.96 mm Diameter, 2 in
Catalog No. 5000527898
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage features a 9.96 mm diameter and a 2-inch overall length. It is designed for precise measurement applications such as setting micrometers, NoGo inspection, and verifying verniers and other measuring devices.
  • Class ZZ Minus tolerance ensures accurate measurements for critical applications.
  • Made from steel for long-lasting performance and reliability.
  • Comes with a Certificate of Accuracy with NIST Traceability, providing assurance of its measurement standards.
  • Ideal for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices.
  • Features a 10 Micro finish for smooth operation and consistent results.
Catalog No. 50-005-27898 Supplier MSC Supplier No. 40437410
This Vermont Gage Class ZZ Minus Plug & Pin Gage features a 9.96 mm diameter and a 2-inch overall length. It is designed for precise measurement applications such as setting micrometers, NoGo inspection, and verifying verniers and other measuring devices.
  • Class ZZ Minus tolerance ensures accurate measurements for critical applications.
  • Made from steel for long-lasting performance and reliability.
  • Comes with a Certificate of Accuracy with NIST Traceability, providing assurance of its measurement standards.
  • Ideal for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices.
  • Features a 10 Micro finish for smooth operation and consistent results.