Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, Steel, Black Oxide
Catalog No. 5000529917
Encompass
This Class ZZ Minus Plug & Pin Gage is engineered for precision measurement tasks, including setting micrometers, NoGo inspection, and integration with verniers and other measuring instruments. It boasts a robust steel construction complemented by a black oxide finish, offering both durability and reliable performance.
  • Features a 0.0440 inch diameter for accurate gauging.
  • Provides a 2 inch overall length for broad application compatibility.
  • Maintains a tight tolerance of 0.000200 inch for consistent results.
  • Supplied with a certificate of accuracy with NIST traceability.
Catalog No. 50-005-29917 Supplier MSC Supplier No. 04709457
This Class ZZ Minus Plug & Pin Gage is engineered for precision measurement tasks, including setting micrometers, NoGo inspection, and integration with verniers and other measuring instruments. It boasts a robust steel construction complemented by a black oxide finish, offering both durability and reliable performance.
  • Features a 0.0440 inch diameter for accurate gauging.
  • Provides a 2 inch overall length for broad application compatibility.
  • Maintains a tight tolerance of 0.000200 inch for consistent results.
  • Supplied with a certificate of accuracy with NIST traceability.