Learn More
MSC Vermont Gage Class ZZ Plug Gage, 0.6930 in, Class ZZ, Tool Steel
Catalog No. 5000531726
Encompass
Class ZZ Plug Gages are economical general-purpose pin gages designed for quick and accurate measurements, including checking hole sizes, calculating distances between holes, gaging slot widths, and checking locations. They can also be used for Go/NoGo gaging and sizing in micrometers and snap gages. This particular gage has a 0.6930 in diameter and is made from tool steel.
  • Tolerance is Class ZZ (.0002 in or .005 mm tolerance)
  • Available as Go (Plus) or NoGo (Minus) gage types
  • NIST traceable
  • 2 in long
  • 10 microinch finish or better
  • Round within .0001 in or .0025 mm
  • Marked with size and direction of tolerance on gages over .0605 in
  • Certificate of Accuracy included
Catalog No. 50-005-31726 Supplier MSC Supplier No. 88969308
Class ZZ Plug Gages are economical general-purpose pin gages designed for quick and accurate measurements, including checking hole sizes, calculating distances between holes, gaging slot widths, and checking locations. They can also be used for Go/NoGo gaging and sizing in micrometers and snap gages. This particular gage has a 0.6930 in diameter and is made from tool steel.
  • Tolerance is Class ZZ (.0002 in or .005 mm tolerance)
  • Available as Go (Plus) or NoGo (Minus) gage types
  • NIST traceable
  • 2 in long
  • 10 microinch finish or better
  • Round within .0001 in or .0025 mm
  • Marked with size and direction of tolerance on gages over .0605 in
  • Certificate of Accuracy included