Learn More
MSC Vermont Gage Class ZZ Plug Gage, .3455 inch, Tool Steel
Catalog No. 5000527387
Encompass
This economical general-purpose plug gage is designed for quick and accurate measurements in various applications. It is ideal for measuring hole sizes, calculating distances between holes, gaging slot widths, and performing Go/NoGo gaging. Built from durable tool steel, this gage provides added wear life and reliable performance.
  • Class ZZ tolerance for precise measurements
  • Constructed from tool steel for durability
  • Standard 2 inch length
  • Features a 10 microinch finish or better
  • Round within .0001 inch
  • NIST traceable
  • Includes certificate of accuracy
Catalog No. 50-005-27387 Supplier MSC Supplier No. 88934559
This economical general-purpose plug gage is designed for quick and accurate measurements in various applications. It is ideal for measuring hole sizes, calculating distances between holes, gaging slot widths, and performing Go/NoGo gaging. Built from durable tool steel, this gage provides added wear life and reliable performance.
  • Class ZZ tolerance for precise measurements
  • Constructed from tool steel for durability
  • Standard 2 inch length
  • Features a 10 microinch finish or better
  • Round within .0001 inch
  • NIST traceable
  • Includes certificate of accuracy