Learn More
MSC Vermont Gage Class ZZ Plug Gage Set, 0.011 to 0.06 inch Range, 50 Pc, Black Oxide Finish
Catalog No. 5000515888
Encompass
This Class ZZ Plug Gage Set includes 50 steel gages with a black oxide finish, providing a diameter range from 0.011 to 0.060 inch and a tolerance of 0.000200 " Each set comes with a Certificate of Accuracy traceable to NIST standards.
  • Includes certificate of accuracy traceable to NIST standards.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Suitable for gaging slot widths, locations, and measuring hole diameters.
  • Ideal for GO gaging inspection and calculating distances between holes.
  • Constructed from durable steel.
  • Features a protective black oxide finish.
  • Contains 50 individual gages.
Catalog No. 50-005-15888 Supplier MSC Supplier No. 84837350
This Class ZZ Plug Gage Set includes 50 steel gages with a black oxide finish, providing a diameter range from 0.011 to 0.060 inch and a tolerance of 0.000200 " Each set comes with a Certificate of Accuracy traceable to NIST standards.
  • Includes certificate of accuracy traceable to NIST standards.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Suitable for gaging slot widths, locations, and measuring hole diameters.
  • Ideal for GO gaging inspection and calculating distances between holes.
  • Constructed from durable steel.
  • Features a protective black oxide finish.
  • Contains 50 individual gages.