Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0265 in Diameter, 2.0000 in OAL, Tool Steel
Catalog No. 5000530744
Encompass
This Class ZZ Plus Plug Gage is ideal for precision Go inspection and setting various measuring devices. It features a 0.0265-inch diameter and a 2.0000-inch overall length, with a tight 0.000200-inch tolerance. Made from durable tool steel, it includes a 10 micro finish and complies with ASME B89.1 standards, ensuring accurate measurements.
  • Precision Go inspection and setting of micrometers and verniers.
  • Diameter of 0.0265 inches.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.000200 inches.
  • Constructed from durable tool steel with a 10 micro finish.
  • Complies with ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-30744 Supplier MSC Supplier No. 88902655
This Class ZZ Plus Plug Gage is ideal for precision Go inspection and setting various measuring devices. It features a 0.0265-inch diameter and a 2.0000-inch overall length, with a tight 0.000200-inch tolerance. Made from durable tool steel, it includes a 10 micro finish and complies with ASME B89.1 standards, ensuring accurate measurements.
  • Precision Go inspection and setting of micrometers and verniers.
  • Diameter of 0.0265 inches.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.000200 inches.
  • Constructed from durable tool steel with a 10 micro finish.
  • Complies with ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.