Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0290 in Diameter, 2.000 in OAL, Tool Steel
Catalog No. 5000516095
Encompass
This plug gage features a 0.0290 inch diameter and a 2.000 inch overall length, made from tool steel with a 10 micro coating/finish. It is designed for "Go" inspection and for setting various measuring devices, ensuring accuracy with a tolerance of 0.000200 "
  • Diameter of 0.0290 "
  • Overall length of 2.000 "
  • Tolerance of 0.000200 "
  • Made from tool steel.
  • 10 micro coating/finish.
  • Suitable for "Go" inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.
Catalog No. 50-005-16095 Supplier MSC Supplier No. 88902903
This plug gage features a 0.0290 inch diameter and a 2.000 inch overall length, made from tool steel with a 10 micro coating/finish. It is designed for "Go" inspection and for setting various measuring devices, ensuring accuracy with a tolerance of 0.000200 "
  • Diameter of 0.0290 "
  • Overall length of 2.000 "
  • Tolerance of 0.000200 "
  • Made from tool steel.
  • 10 micro coating/finish.
  • Suitable for "Go" inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.