Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0440 in Diameter, 2.0000 in OAL
Catalog No. 5000511123
Encompass
This Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 0.0440 inch diameter and a 2.0000 inch overall length, making it suitable for go inspection and setting various measuring devices.
  • Has a 0.000200 in tolerance.
  • Features a black oxide coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from durable tool steel.
  • Ideal for go inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Complies with ASME B89.1 standards.
Catalog No. 50-005-11123 Supplier MSC Supplier No. 03529120
This Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 0.0440 inch diameter and a 2.0000 inch overall length, making it suitable for go inspection and setting various measuring devices.
  • Has a 0.000200 in tolerance.
  • Features a black oxide coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from durable tool steel.
  • Ideal for go inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Complies with ASME B89.1 standards.