Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0515 in Diameter, 0.0002 in Tolerance
Catalog No. 5000516099
Encompass
This Class ZZ Plus Plug Gage provides precise measurement for quality control applications. Engineered to stringent specifications, it is ideal for verifying hole sizes and ensuring manufacturing accuracy in various industrial settings.
  • Precision ground to Class ZZ specifications
  • Diameter of 0.0515 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • 10 microinch surface finish
Catalog No. 50-005-16099 Supplier MSC Supplier No. 88905153
This Class ZZ Plus Plug Gage provides precise measurement for quality control applications. Engineered to stringent specifications, it is ideal for verifying hole sizes and ensuring manufacturing accuracy in various industrial settings.
  • Precision ground to Class ZZ specifications
  • Diameter of 0.0515 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • 10 microinch surface finish