Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0530 in Diameter, 2.0000 in Length, Tool Steel
Catalog No. 5000534931
Encompass
This Class ZZ Plus Plug Gage has a 0.0530 in diameter and a 2.0000 in overall length, featuring a 0.000200 in tolerance and a black oxide finish. It is used for "Go" inspection and for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
  • Constructed from tool steel
Catalog No. 50-005-34931 Supplier MSC Supplier No. 50734045
This Class ZZ Plus Plug Gage has a 0.0530 in diameter and a 2.0000 in overall length, featuring a 0.000200 in tolerance and a black oxide finish. It is used for "Go" inspection and for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
  • Constructed from tool steel