Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0645 in Dia, 0.000200 in Tolerance, Tool Steel
Catalog No. 5000531710
Encompass
This Class ZZ Plus Plug Gage is engineered for precise measurement and inspection applications. Constructed from durable tool steel, it features a 0.0645 inch diameter and a tight tolerance of 0.000200 " It is ideal for Go inspection, setting micrometers, verniers, and other measuring devices, and comes with a certificate of accuracy traceable to NIST.
  • 0.000200 inch tolerance
  • 10 micro coating/finish
  • Includes certificate of accuracy with NIST traceability
  • Made from tool steel
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Conforms to ASME B89.1 standards
Catalog No. 50-005-31710 Supplier MSC Supplier No. 88906458
This Class ZZ Plus Plug Gage is engineered for precise measurement and inspection applications. Constructed from durable tool steel, it features a 0.0645 inch diameter and a tight tolerance of 0.000200 " It is ideal for Go inspection, setting micrometers, verniers, and other measuring devices, and comes with a certificate of accuracy traceable to NIST.
  • 0.000200 inch tolerance
  • 10 micro coating/finish
  • Includes certificate of accuracy with NIST traceability
  • Made from tool steel
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Conforms to ASME B89.1 standards