Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0650 Dia, 2.0000 OAL
Catalog No. 5000516128
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications. Made from durable tool steel, it features a 0.0650 in diameter and 2.0000 in overall length, suitable for various inspection and calibration tasks.
  • For go inspection, setting micrometers, verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • 10 micro coating/finish
  • Constructed from durable tool steel
  • 0.0650 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance
Catalog No. 50-005-16128 Supplier MSC Supplier No. 88906508
This Class ZZ Plus plug gage is designed for precise measurement applications. Made from durable tool steel, it features a 0.0650 in diameter and 2.0000 in overall length, suitable for various inspection and calibration tasks.
  • For go inspection, setting micrometers, verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • 10 micro coating/finish
  • Constructed from durable tool steel
  • 0.0650 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance