Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0670 In Dia, 2.0000 In OAL, Tool Steel
Catalog No. 5000531711
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for precision Go inspection, micrometer setting, vernier calibration, and other measuring devices. It features a 0.0670 inch diameter and a 2.0000 inch overall length, crafted from durable tool steel.
  • Designed for Go inspection, setting micrometers, verniers, and other measuring devices.
  • Manufactured from durable tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Features a 10 micro coating/finish.
  • Provides a tolerance of 0.000200 "
Catalog No. 50-005-31711 Supplier MSC Supplier No. 88906706
This Vermont Gage Class ZZ Plus Plug Gage is designed for precision Go inspection, micrometer setting, vernier calibration, and other measuring devices. It features a 0.0670 inch diameter and a 2.0000 inch overall length, crafted from durable tool steel.
  • Designed for Go inspection, setting micrometers, verniers, and other measuring devices.
  • Manufactured from durable tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Features a 10 micro coating/finish.
  • Provides a tolerance of 0.000200 "