Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0790 in Dia, 2.0000 in OAL
Catalog No. 5000516439
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate go inspection and setting of various measuring devices. Constructed from durable tool steel, this gage features a diameter of 0.0790 in and an overall length of 2.0000 in.
  • Tolerance of 0.000200 in
  • 10 micro coating and finish
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Made in the United States
Catalog No. 50-005-16439 Supplier MSC Supplier No. 88907902
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate go inspection and setting of various measuring devices. Constructed from durable tool steel, this gage features a diameter of 0.0790 in and an overall length of 2.0000 in.
  • Tolerance of 0.000200 in
  • 10 micro coating and finish
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Made in the United States