Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0815 in, 2.0000 in OAL, Tool steel
Catalog No. 5000535700
Encompass
The Class ZZ Plus Plug Gage is a precision measuring tool with a diameter of 0.0815 in and an overall length of 2.0000 in. It has a tolerance of 0.000200 in and a 10 micro coating/finish. This plug gage is made of tool steel and is designed for "Go" inspection, as well as for setting micrometers, verniers, and other measuring devices. It includes a Certificate of Accuracy with NIST Traceability.
  • Material: Tool steel
  • Application for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Made in the United States
Catalog No. 50-005-35700 Supplier MSC Supplier No. 88908157
The Class ZZ Plus Plug Gage is a precision measuring tool with a diameter of 0.0815 in and an overall length of 2.0000 in. It has a tolerance of 0.000200 in and a 10 micro coating/finish. This plug gage is made of tool steel and is designed for "Go" inspection, as well as for setting micrometers, verniers, and other measuring devices. It includes a Certificate of Accuracy with NIST Traceability.
  • Material: Tool steel
  • Application for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Made in the United States