Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0820" Dia, 2.0000" OAL, Tool Steel
Catalog No. 5000511319
Encompass
The Class ZZ Plus Plug Gage is designed for precise 'Go' inspection and for calibrating various measuring instruments. This durable gage is crafted from tool steel to ensure accuracy and longevity in demanding laboratory and industrial environments.
  • Designed for 'Go' inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Constructed from tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 0.000200" tolerance.
Catalog No. 50-005-11319 Supplier MSC Supplier No. 88908207
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Class ZZ Plus Plug Gage is designed for precise 'Go' inspection and for calibrating various measuring instruments. This durable gage is crafted from tool steel to ensure accuracy and longevity in demanding laboratory and industrial environments.
  • Designed for 'Go' inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Constructed from tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 0.000200" tolerance.