Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0845 in Diameter, Tool Steel
Catalog No. 5000530844
Encompass
This Class ZZ Plus Plug Gage is designed for go inspection and setting various measuring devices such as micrometers and verniers. It features a precise diameter of 0.0845 inches and an overall length of 2.0000 inches, manufactured from durable tool steel.
  • Designed for go inspection and setting measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from durable tool steel
  • Features a 10 micro finish
  • Complies with ASME B89.1 standards
  • Offers a precise measurement diameter
  • Provides a consistent overall length
  • Ensures tight tolerance for accurate measurements
Catalog No. 50-005-30844 Supplier MSC Supplier No. 88908454
This Class ZZ Plus Plug Gage is designed for go inspection and setting various measuring devices such as micrometers and verniers. It features a precise diameter of 0.0845 inches and an overall length of 2.0000 inches, manufactured from durable tool steel.
  • Designed for go inspection and setting measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from durable tool steel
  • Features a 10 micro finish
  • Complies with ASME B89.1 standards
  • Offers a precise measurement diameter
  • Provides a consistent overall length
  • Ensures tight tolerance for accurate measurements