Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0885 in, Tool Steel
Catalog No. 5000535778
Encompass
This Class ZZ Plus plug gage is engineered for precise 'Go' inspection and calibrating measuring devices like micrometers and verniers. Manufactured from durable tool steel, it offers a 0.0885 inch diameter and a precise 0.000200 inch tolerance, making it suitable for critical measurement applications.
  • Designed for go inspection and setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Constructed from durable tool steel.
  • Features a 0.0885 inch diameter and 2.0000 inch overall length.
  • Provides a precise 0.000200 inch tolerance.
  • Finished with a 10 micro coating.
Catalog No. 50-005-35778 Supplier MSC Supplier No. 88908850
This Class ZZ Plus plug gage is engineered for precise 'Go' inspection and calibrating measuring devices like micrometers and verniers. Manufactured from durable tool steel, it offers a 0.0885 inch diameter and a precise 0.000200 inch tolerance, making it suitable for critical measurement applications.
  • Designed for go inspection and setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Constructed from durable tool steel.
  • Features a 0.0885 inch diameter and 2.0000 inch overall length.
  • Provides a precise 0.000200 inch tolerance.
  • Finished with a 10 micro coating.