Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.0920 in Dia, 2.0000 in OAL
Catalog No. 5000529442
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurements and calibration in industrial applications. It features a 0.0920 inch diameter and a 2.0000 inch overall length, constructed from durable tool steel with a black oxide finish for enhanced performance.
  • Used for go inspection
  • Sets micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Tolerance of 0.000200 in
  • Made from tool steel
  • Features a black oxide finish
Catalog No. 50-005-29442 Supplier MSC Supplier No. 03529542
This Class ZZ Plus Plug Gage is designed for precise measurements and calibration in industrial applications. It features a 0.0920 inch diameter and a 2.0000 inch overall length, constructed from durable tool steel with a black oxide finish for enhanced performance.
  • Used for go inspection
  • Sets micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Tolerance of 0.000200 in
  • Made from tool steel
  • Features a black oxide finish