Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1100 in Diameter, 2.0000 in OAL, Tool Steel
Catalog No. 5000535485
Encompass
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection, as well as setting micrometers, verniers, and other measuring devices. Manufactured from durable tool steel, it ensures reliability and accuracy for various measurement applications.
  • Diameter of 0.1100 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • Constructed from tool steel
  • Features a 10 micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-35485 Supplier MSC Supplier No. 88911003
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection, as well as setting micrometers, verniers, and other measuring devices. Manufactured from durable tool steel, it ensures reliability and accuracy for various measurement applications.
  • Diameter of 0.1100 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • Constructed from tool steel
  • Features a 10 micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards