Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1150 in, 2.0000 in, Tool Steel
Catalog No. 5000516325
Encompass
This Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a diameter of 0.1150 in and an overall length of 2.0000 in, crafted from durable tool steel. It is suitable for "Go" inspection and for setting micrometers, verniers, and other measuring devices, ensuring accuracy and reliability.
  • 0.1150 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • Made from tool steel
  • Conforms to ASME B89.1 standards
  • Ideal for go inspection
  • Useful for setting micrometers, verniers, and other measuring devices
  • 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-16325 Supplier MSC Supplier No. 88911508
This Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a diameter of 0.1150 in and an overall length of 2.0000 in, crafted from durable tool steel. It is suitable for "Go" inspection and for setting micrometers, verniers, and other measuring devices, ensuring accuracy and reliability.
  • 0.1150 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • Made from tool steel
  • Conforms to ASME B89.1 standards
  • Ideal for go inspection
  • Useful for setting micrometers, verniers, and other measuring devices
  • 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability