Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1155 in, Tool Steel, 10 Micro Finish
Catalog No. 5000531688
Encompass
This plug gage is engineered for high-precision go inspection and for accurately setting various measuring devices such as micrometers and verniers. Manufactured from durable tool steel, this gage features a specific diameter, overall length, and tight tolerance to ensure reliability and accuracy in critical measurements.
  • Precisely measures 0.1155 inch diameter
  • Constructed from durable tool steel for longevity
  • Features a 10 micro inch finish for smooth operation
  • Meets ASME B89.1 standards for quality assurance
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for go inspection and calibrating measuring instruments
Catalog No. 50-005-31688 Supplier MSC Supplier No. 88911557
This plug gage is engineered for high-precision go inspection and for accurately setting various measuring devices such as micrometers and verniers. Manufactured from durable tool steel, this gage features a specific diameter, overall length, and tight tolerance to ensure reliability and accuracy in critical measurements.
  • Precisely measures 0.1155 inch diameter
  • Constructed from durable tool steel for longevity
  • Features a 10 micro inch finish for smooth operation
  • Meets ASME B89.1 standards for quality assurance
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for go inspection and calibrating measuring instruments