Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1160 in Diameter, 2.0000 in OAL, Tool Steel
Catalog No. 5000531689
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for precision 'Go' inspection and for accurately setting micrometers, verniers, and other measuring devices. This plug gage has a diameter of 0.1160 inch and an overall length of 2.0000 inch, featuring a robust tool steel construction with a 10 Micro finish for durability and accuracy. It adheres to ASME B89.1 standards and comes with a Certificate of Accuracy that is traceable to NIST.
  • Designed for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Features a diameter of 0.1160 inch
  • Provides an overall length of 2.0000 inch
  • Maintains a tolerance of 0.000200 inch
  • Constructed from durable tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Features a 10 micro finish
Catalog No. 50-005-31689 Supplier MSC Supplier No. 88911607
The Vermont Gage Class ZZ Plus Plug Gage is designed for precision 'Go' inspection and for accurately setting micrometers, verniers, and other measuring devices. This plug gage has a diameter of 0.1160 inch and an overall length of 2.0000 inch, featuring a robust tool steel construction with a 10 Micro finish for durability and accuracy. It adheres to ASME B89.1 standards and comes with a Certificate of Accuracy that is traceable to NIST.
  • Designed for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Features a diameter of 0.1160 inch
  • Provides an overall length of 2.0000 inch
  • Maintains a tolerance of 0.000200 inch
  • Constructed from durable tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Features a 10 micro finish