Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1290 in Diameter, 2.0000 in Length
Catalog No. 5000533454
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection. It features a 0.1290 inch diameter and a 2.0000 inch overall length, making it suitable for various metrology applications.
  • Manufactured from durable tool steel
  • Black oxide coating for protection
  • 0.000200 inch tolerance for accuracy
  • Suitable for go inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Made in the United States
Catalog No. 50-005-33454 Supplier MSC Supplier No. 03529971
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection. It features a 0.1290 inch diameter and a 2.0000 inch overall length, making it suitable for various metrology applications.
  • Manufactured from durable tool steel
  • Black oxide coating for protection
  • 0.000200 inch tolerance for accuracy
  • Suitable for go inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Made in the United States