Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1330 in, 0.000200 in
Catalog No. 5000529030
Encompass
This Class ZZ Plus Plug Gage features a 0.1330 inch diameter and a 2.0000 inch overall length, with a 0.000200 inch tolerance and 10 micro finish. Constructed from tool steel, it is designed for "Go" inspection and for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability and meets ASME B89.1 standards.
  • 0.1330 inch diameter for precise measurements.
  • 2.0000 inch overall length.
  • 0.000200 inch tolerance ensures accuracy.
  • Constructed from durable tool steel.
  • Designed for "Go" inspection applications.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Features a 10 micro finish.
Catalog No. 50-005-29030 Supplier MSC Supplier No. 88913306
This Class ZZ Plus Plug Gage features a 0.1330 inch diameter and a 2.0000 inch overall length, with a 0.000200 inch tolerance and 10 micro finish. Constructed from tool steel, it is designed for "Go" inspection and for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability and meets ASME B89.1 standards.
  • 0.1330 inch diameter for precise measurements.
  • 2.0000 inch overall length.
  • 0.000200 inch tolerance ensures accuracy.
  • Constructed from durable tool steel.
  • Designed for "Go" inspection applications.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Features a 10 micro finish.