Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1360 in, Tool Steel, Black Oxide
Catalog No. 5000529677
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and calibration in industrial applications. It is ideal for go inspection and for setting micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a black oxide coating for enhanced protection.
  • Maintains a precision tolerance of 0.000200 in.
  • Overall length of 2.0000 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
Catalog No. 50-005-29677 Supplier MSC Supplier No. 05786165
This Class ZZ Plus plug gage is designed for precise measurement and calibration in industrial applications. It is ideal for go inspection and for setting micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a black oxide coating for enhanced protection.
  • Maintains a precision tolerance of 0.000200 in.
  • Overall length of 2.0000 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.