Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1410 in, 0.000200 in Tolerance
Catalog No. 5000535751
Encompass
This Class ZZ Plus Plug Gage is designed for precise Go Inspection, setting micrometers, verniers, and other measuring devices. It features a 0.1410 inch diameter, 2.0000 inch overall length, and a tight 0.000200 inch tolerance, making it suitable for demanding laboratory and inspection applications.
  • Precise 0.1410 inch diameter for accurate measurements.
  • 2.0000 inch overall length for versatility.
  • Tight 0.000200 inch tolerance ensures high precision.
  • Constructed from durable tool steel.
  • Ideal for Go Inspection and setting various measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-35751 Supplier MSC Supplier No. 88914106
This Class ZZ Plus Plug Gage is designed for precise Go Inspection, setting micrometers, verniers, and other measuring devices. It features a 0.1410 inch diameter, 2.0000 inch overall length, and a tight 0.000200 inch tolerance, making it suitable for demanding laboratory and inspection applications.
  • Precise 0.1410 inch diameter for accurate measurements.
  • 2.0000 inch overall length for versatility.
  • Tight 0.000200 inch tolerance ensures high precision.
  • Constructed from durable tool steel.
  • Ideal for Go Inspection and setting various measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 micro coating/finish.