Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1420 in Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000535703
Encompass
This Class ZZ Plus Plug Gage features a 0.1420 in diameter and 2.0000 in overall length, constructed from tool steel with a 10 micro finish. Adhering to ASME B89.1 standards, this gage is designed for Go Inspection and for setting various measuring devices, and includes a Certificate of Accuracy with NIST Traceability.
  • Features a 0.1420 in diameter and 2.0000 in overall length.
  • Constructed from durable tool steel for longevity.
  • Smooth 10 micro finish ensures accuracy.
  • Meets ASME B89.1 standards for reliable performance.
  • Ideal for go inspection.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Maintains a tight tolerance of 0.000200 in.
Catalog No. 50-005-35703 Supplier MSC Supplier No. 88914205
This Class ZZ Plus Plug Gage features a 0.1420 in diameter and 2.0000 in overall length, constructed from tool steel with a 10 micro finish. Adhering to ASME B89.1 standards, this gage is designed for Go Inspection and for setting various measuring devices, and includes a Certificate of Accuracy with NIST Traceability.
  • Features a 0.1420 in diameter and 2.0000 in overall length.
  • Constructed from durable tool steel for longevity.
  • Smooth 10 micro finish ensures accuracy.
  • Meets ASME B89.1 standards for reliable performance.
  • Ideal for go inspection.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Maintains a tight tolerance of 0.000200 in.