Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1490 in, Tool Steel, 2 in
Catalog No. 5000530848
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is an economical, general-purpose pin gage designed for quick and accurate measurements. It is suitable for various applications, including measuring hole diameters, calculating distances between holes, gaging slot widths, checking locations, and Go/NoGo inspection. This 0.1490 inch diameter gage is made of durable tool steel and is 2 inches long, ensuring reliable performance in laboratory and industrial settings.
  • Made from tool steel with a 60/65 Rc hardness.
  • Features Class ZZ tolerance of 0.0002 "
  • Measures 2 inches in overall length.
  • Includes a 10 microinch finish or better.
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices.
  • Comes with a certificate of accuracy, traceable to NIST.
  • Complies with ASME B89.1 standards.
Catalog No. 50-005-30848 Supplier MSC Supplier No. 88914908
The Vermont Gage Class ZZ Plus Plug Gage is an economical, general-purpose pin gage designed for quick and accurate measurements. It is suitable for various applications, including measuring hole diameters, calculating distances between holes, gaging slot widths, checking locations, and Go/NoGo inspection. This 0.1490 inch diameter gage is made of durable tool steel and is 2 inches long, ensuring reliable performance in laboratory and industrial settings.
  • Made from tool steel with a 60/65 Rc hardness.
  • Features Class ZZ tolerance of 0.0002 "
  • Measures 2 inches in overall length.
  • Includes a 10 microinch finish or better.
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices.
  • Comes with a certificate of accuracy, traceable to NIST.
  • Complies with ASME B89.1 standards.