Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1560 in Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000535780
Encompass
This plug gage is designed for precise 'Go' inspection and calibrating measuring devices. It features a 0.1560 in diameter and 2.0000 in overall length, is constructed from durable tool steel, and meets ASME B89.1 standards with a 10 micro-inch finish.
  • For 'Go' inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Constructed from tool steel
  • Meets ASME B89.1 standards
  • Features a 10 micro-inch finish
Catalog No. 50-005-35780 Supplier MSC Supplier No. 88915608
This plug gage is designed for precise 'Go' inspection and calibrating measuring devices. It features a 0.1560 in diameter and 2.0000 in overall length, is constructed from durable tool steel, and meets ASME B89.1 standards with a 10 micro-inch finish.
  • For 'Go' inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Constructed from tool steel
  • Meets ASME B89.1 standards
  • Features a 10 micro-inch finish