Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1595 in, 0.0002 in, 10 Microfinish
Catalog No. 5000530506
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel with a 10 microfinish, this gage features a 0.1595 inch diameter and a 0.0002 inch tolerance, ensuring high accuracy for various applications.
  • For Go inspection, set micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Constructed from tool steel.
  • Features a 10 microfinish.
Catalog No. 50-005-30506 Supplier MSC Supplier No. 88915954
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel with a 10 microfinish, this gage features a 0.1595 inch diameter and a 0.0002 inch tolerance, ensuring high accuracy for various applications.
  • For Go inspection, set micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Constructed from tool steel.
  • Features a 10 microfinish.