Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1900 in Dia, 2.0000 in OAL
Catalog No. 5000527984
Encompass
This plug gage is designed for precise measurement and inspection applications. It features a 0.1900 inch diameter and a 2.0000 inch overall length, with a tight tolerance of 0.000200 " Made from durable tool steel and finished with black oxide, it meets ASME B89.1 standards.
  • Certificate of accuracy with NIST traceability
  • For go inspection; setting micrometers, verniers, and other measuring devices
  • Manufactured from tool steel
  • Black oxide coating/finish
  • Meets ASME B89.1 standards
Catalog No. 50-005-27984 Supplier MSC Supplier No. 74235599
This plug gage is designed for precise measurement and inspection applications. It features a 0.1900 inch diameter and a 2.0000 inch overall length, with a tight tolerance of 0.000200 " Made from durable tool steel and finished with black oxide, it meets ASME B89.1 standards.
  • Certificate of accuracy with NIST traceability
  • For go inspection; setting micrometers, verniers, and other measuring devices
  • Manufactured from tool steel
  • Black oxide coating/finish
  • Meets ASME B89.1 standards