Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.1945 in, 0.000200 in Tolerance
Catalog No. 5000535753
Encompass
This plug gage is designed for Go Inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.1945 in diameter and a 2.0000 in overall length. The gage is constructed from tool steel and adheres to ASME B89.1 standards.
  • Class ZZ Plus gage
  • 0.000200 in tolerance
  • 10 Micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Constructed from tool steel
  • Adheres to ASME B89.1 standards
Catalog No. 50-005-35753 Supplier MSC Supplier No. 88919451
This plug gage is designed for Go Inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.1945 in diameter and a 2.0000 in overall length. The gage is constructed from tool steel and adheres to ASME B89.1 standards.
  • Class ZZ Plus gage
  • 0.000200 in tolerance
  • 10 Micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
  • Constructed from tool steel
  • Adheres to ASME B89.1 standards