Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2015 in, Tool Steel
Catalog No. 5000511294
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection applications. Crafted from durable tool steel, this plug gage features a 0.2015 inch diameter and a 2.0000 inch overall length, with a tight tolerance of 0.000200 " It is suitable for 'Go' inspection and for setting micrometers, verniers, and other measuring devices.
  • 0.000200 inch tolerance
  • 10 micro coating/finish
  • Certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Suitable for 'go' inspection and setting measuring devices
  • Hassle-free guarantee for exchanges or refunds
Catalog No. 50-005-11294 Supplier MSC Supplier No. 88920152
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection applications. Crafted from durable tool steel, this plug gage features a 0.2015 inch diameter and a 2.0000 inch overall length, with a tight tolerance of 0.000200 " It is suitable for 'Go' inspection and for setting micrometers, verniers, and other measuring devices.
  • 0.000200 inch tolerance
  • 10 micro coating/finish
  • Certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Suitable for 'go' inspection and setting measuring devices
  • Hassle-free guarantee for exchanges or refunds